Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024129.1 | NP_077043.1 | 6 - 7 | 777 | 65 | |
XM_006241285.1 | XP_006241347.1 | 6 - 7 | 962 | 65 | ||
GenBank mRNA | BC083750.1 | - | 6 - 7 | 777 | 7192499 | 65 |
FQ214818.1 | - | 4 - 5 | 436 | 65 | ||
FQ219808.1 | - | 6 - 7 | 810 | 65 | ||
FQ220209.1 | - | 6 - 7 | 843 | 65 | ||
FQ220915.1 | - | 2 - 3 | 130 | 65 | ||
FQ222189.1 | - | 6 - 7 | 846 | 65 | ||
FQ223165.1 | - | 2 - 3 | 217 | 65 | ||
FQ223184.1 | - | 6 - 7 | 851 | 65 | ||
FQ223539.1 | - | 1 - 2 | 69 | 65 | ||
FQ228702.1 | - | 6 - 7 | 801 | 65 | ||
FQ229041.1 | - | 6 - 7 | 865 | 65 | ||
FQ229786.1 | - | 6 - 7 | 846 | 65 | ||
X59859.1 | - | 5 - 6 | 697 | 65 | ||
Z12298.1 | - | 6 - 7 | 906 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |