Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ly6h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134839.1 | NP_001128311.1 | 2 - 3 | 213 | 85 | |
XM_006241781.2 | XP_006241843.1 | 2 - 3 | 237 | 85 | ||
XM_006241782.2 | XP_006241844.1 | 3 - 4 | 423 | 85 | ||
XM_006241783.2 | XP_006241845.1 | 2 - 3 | 162 | 85 | ||
GenBank mRNA | BC169114.1 | - | 2 - 3 | 195 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |