Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031039.1 | NP_112301.1 | 9 - 10 | 1397 | 96 | |
XM_008765599.1 | XP_008763821.1 | 10 - 11 | 2117 | 96 | ||
GenBank mRNA | BC085728.1 | - | 4 - 5 | 1882 | 96 | |
BC097937.1 | - | 9 - 10 | 1401 | 7324977 | 96 | |
D10354.1 | - | 9 - 10 | 1397 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |