Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134529.1 | NP_001128001.1 | 4 - 5 | 556 | 92 | |
XM_006241976.2 | XP_006242038.1 | 4 - 5 | 702 | 92 | ||
XM_006241977.2 | XP_006242039.1 | 4 - 5 | 696 | 92 | ||
XM_006241978.2 | XP_006242040.1 | 4 - 5 | 699 | 92 | ||
XM_006241979.2 | XP_006242041.1 | 4 - 5 | 696 | 92 | ||
XM_006241981.2 | XP_006242043.1 | 4 - 5 | 693 | 92 | ||
XM_006241982.2 | XP_006242044.1 | 4 - 5 | 692 | 92 | ||
XM_006241983.2 | XP_006242045.1 | 4 - 5 | 692 | 92 | ||
XM_008765708.1 | XP_008763930.1 | 4 - 5 | 699 | 92 | ||
XM_008765709.1 | XP_008763931.1 | 4 - 5 | 697 | 92 | ||
XM_008765710.1 | XP_008763932.1 | 4 - 5 | 700 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Ion Channel 3' Most Rattus norvegicus Probe spans exons |