Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam91a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127578.1 | NP_001121050.1 | 12 - 13 | 1302 | 110 | |
XM_006241684.2 | XP_006241746.1 | 12 - 13 | 1306 | 110 | ||
GenBank mRNA | BC166852.1 | - | 12 - 13 | 1302 | 9092514 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |