Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006985.1 | NP_001006986.1 | 4 - 5 | 304 | 61 | |
XM_006241651.1 | XP_006241713.1 | 4 - 5 | 499 | 61 | ||
GenBank mRNA | BC084710.1 | - | 4 - 5 | 304 | 7302342 | 61 |
FQ220388.1 | - | 4 - 5 | 485 | 61 | ||
FQ232270.1 | - | 4 - 5 | 348 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |