Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130493.1 | NP_001123965.1 | 2 - 3 | 385 | 99 | |
XM_008765565.1 | XP_008763787.1 | 2 - 3 | 622 | 99 | ||
GenBank mRNA | BC158731.1 | - | 2 - 3 | 295 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons |