Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyth4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130577.1 | NP_001124049.1 | 7 - 8 | 718 | 83 | |
XM_006242031.2 | XP_006242093.1 | 8 - 9 | 805 | 83 | ||
XM_006242033.2 | XP_006242095.1 | 7 - 8 | 722 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |