Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033231.1 | NP_150234.1 | 7 - 8 | 1206 | 67 | |
XM_006230105.2 | XP_006230167.1 | 7 - 8 | 1225 | 67 | ||
XM_006230106.2 | XP_006230168.1 | 7 - 8 | 1186 | 67 | ||
GenBank mRNA | BC090325.1 | - | 7 - 8 | 1206 | 7128525 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |