Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crym | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053955.1 | NP_446407.1 | 1 - 2 | 205 | 114 | |
GenBank mRNA | BC088121.1 | - | 1 - 2 | 236 | 7323247 | 114 |
Y17328.1 | - | 1 - 2 | 205 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |