Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hpx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053318.1 | NP_445770.1 | 2 - 3 | 164 | 69 | |
GenBank mRNA | BC091137.1 | - | 2 - 3 | 155 | 7326169 | 69 |
FQ210231.1 | - | 2 - 3 | 165 | 69 | ||
FQ211176.1 | - | 2 - 3 | 165 | 69 | ||
FQ218633.1 | - | 2 - 3 | 163 | 69 | ||
FQ218971.1 | - | 2 - 3 | 157 | 69 | ||
FQ219309.1 | - | 2 - 3 | 163 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |