Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hpx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053318.1 | NP_445770.1 | 5 - 6 | 502 | 76 | |
GenBank mRNA | BC091137.1 | - | 5 - 6 | 493 | 7326169 | 76 |
FQ210231.1 | - | 5 - 6 | 503 | 76 | ||
FQ211176.1 | - | 5 - 6 | 503 | 76 | ||
FQ218633.1 | - | 5 - 6 | 501 | 76 | ||
FQ218971.1 | - | 5 - 6 | 495 | 76 | ||
FQ218998.1 | - | 2 - 3 | 271 | 76 | ||
FQ219309.1 | - | 5 - 6 | 501 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |