Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012460.1 | NP_001012478.1 | 5 - 6 | 483 | 68 | |
GenBank mRNA | BC089897.1 | - | 5 - 6 | 483 | 7381516 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |