Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198736.1 | NP_942031.1 | 2 - 3 | 588 | 64 | |
GenBank mRNA | AY305029.1 | - | 2 - 3 | 588 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Rattus norvegicus Probe spans exons |