Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmtc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135858.1 | NP_001129330.1 | 11 - 12 | 1719 | 68 | |
XM_008765316.1 | XP_008763538.1 | 10 - 11 | 1629 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |