Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002729781.3 | XP_002729827.2 | 3 - 4 | 340 | 77 | |
XM_008765341.1 | XP_008763563.1 | 3 - 4 | 505 | 77 | ||
XM_008765342.1 | XP_008763564.1 | 2 - 3 | 572 | 77 | ||
GenBank mRNA | FQ231893.1 | - | 3 - 4 | 404 | 77 | |
FQ233318.1 | - | 3 - 4 | 404 | 77 | ||
FQ235303.1 | - | 3 - 4 | 404 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |