Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pawr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033485.2 | NP_277020.1 | 1 - 2 | 557 | 64 | |
XM_006241317.2 | XP_006241379.1 | 2 - 3 | 770 | 64 | ||
GenBank mRNA | U05989.1 | - | 1 - 2 | 557 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |