Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prp2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768296.1 | XP_008766518.1 | 6 - 7 | 12584 | 134 | |
GenBank mRNA | BC101875.1 | - | 3 - 4 | 198 | 134 | |
M86514.1 | - | 15957 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Rattus norvegicus Probe spans exons |