Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014162.4 | NP_001014184.3 | 3 - 4 | 664 | 67 | |
XM_006230123.1 | XP_006230185.1 | 4 - 5 | 607 | 67 | ||
XM_006230126.1 | XP_006230188.1 | 4 - 5 | 607 | 67 | ||
GenBank mRNA | AY325244.1 | - | 900 | 67 | ||
BC078915.1 | - | 3 - 4 | 663 | 7096091 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |