Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014022.1 | NP_001014044.1 | 6 - 7 | 885 | 128 | |
XM_006230033.2 | XP_006230095.1 | 5 - 6 | 2340 | 128 | ||
XM_006230034.2 | XP_006230096.1 | 8 - 9 | 1365 | 128 | ||
XM_006230035.2 | XP_006230097.1 | 7 - 8 | 1336 | 128 | ||
XM_006230036.2 | XP_006230098.1 | 7 - 8 | 1300 | 128 | ||
XM_006230038.2 | XP_006230100.1 | 6 - 7 | 883 | 128 | ||
XM_006230040.2 | XP_006230102.1 | 5 - 6 | 1159 | 128 | ||
XM_006230041.2 | XP_006230103.1 | 7 - 8 | 1207 | 128 | ||
XM_006230043.2 | XP_006230105.1 | 5 - 6 | 3466 | 128 | ||
XM_008759724.1 | XP_008757946.1 | 6 - 7 | 2425 | 128 | ||
XM_008759725.1 | XP_008757947.1 | 6 - 7 | 1255 | 128 | ||
XM_008759726.1 | XP_008757948.1 | 5 - 6 | 2340 | 128 | ||
GenBank mRNA | BC090330.1 | - | 6 - 7 | 885 | 7300759 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |