Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006229698.2 | XP_006229760.1 | 3 - 4 | 559 | 76 | |
XM_006229699.2 | XP_006229761.1 | 3 - 4 | 890 | 76 | ||
XM_008759650.1 | XP_008757872.1 | 3 - 4 | 311 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus Probe spans exons |