Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109156.1 | NP_001102626.1 | 4 - 5 | 667 | 130 | |
XM_006230324.2 | XP_006230386.1 | 4 - 5 | 1338 | 130 | ||
GenBank mRNA | BC161977.1 | - | 4 - 5 | 630 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |