Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfeb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025707.1 | NP_001020878.1 | 1 - 2 | 459 | 104 | |
XM_006244440.2 | XP_006244502.1 | 2 - 3 | 1388 | 104 | ||
XM_006244442.1 | XP_006244504.1 | 2 - 3 | 378 | 104 | ||
XM_006244446.2 | XP_006244508.1 | 2 - 3 | 1649 | 104 | ||
GenBank mRNA | BC099102.1 | - | 1 - 2 | 459 | 7460021 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |