Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bai3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106898.1 | NP_001100368.1 | 11 - 12 | 2206 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Rattus norvegicus Probe spans exons |