Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bai3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106898.1 | NP_001100368.1 | 20 - 21 | 3177 | 73 | |
XM_008766928.1 | XP_008765150.1 | 4 - 5 | 868 | 73 | ||
XM_008766929.1 | XP_008765151.1 | 5 - 6 | 761 | 73 | ||
XM_008766930.1 | XP_008765152.1 | 5 - 6 | 2182 | 73 | ||
XM_008766931.1 | XP_008765153.1 | 5 - 6 | 998 | 73 | ||
XM_008766932.1 | XP_008765154.1 | 4 - 5 | 868 | 73 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Rattus norvegicus Probe spans exons |