Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006244436.2 | XP_006244498.1 | 2 - 3 | 556 | 111 | |
XM_006244437.2 | XP_006244499.1 | 2 - 3 | 410 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |