Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mocs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106881.1 | NP_001100351.1 | 8 - 9 | 1055 | 62 | |
XM_006244415.2 | XP_006244477.1 | 9 - 10 | 1125 | 62 | ||
XM_006244416.2 | XP_006244478.1 | 8 - 9 | 1066 | 62 | ||
XM_006244418.2 | XP_006244480.1 | 9 - 10 | 1120 | 62 | ||
XM_008766840.1 | XP_008765062.1 | 5 - 6 | 473 | 62 | ||
XM_008766841.1 | XP_008765063.1 | 8 - 9 | 1064 | 62 | ||
XM_008766842.1 | XP_008765064.1 | 3 - 4 | 367 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |