Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750667.3 | XP_003750715.3 | 5 - 6 | 718 | 70 | |
XM_003750668.3 | XP_003750716.3 | 6 - 7 | 800 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |