Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubox5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033997.1 | NP_001029169.1 | 3 - 4 | 361 | 98 | |
XM_008762160.1 | XP_008760382.1 | 2 - 3 | 272 | 98 | ||
XM_008762161.1 | XP_008760383.1 | 2 - 3 | 272 | 98 | ||
GenBank mRNA | BC101912.1 | - | 3 - 4 | 361 | 7933511 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |