Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtch2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106488.2 | NP_001099958.2 | 703 | 98 | ||
GenBank mRNA | BC158687.1 | - | 654 | 8375010 | 98 | |
CK366546.1 | - | 100 | 98 | |||
EV779017.1 | - | 724 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |