Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Traf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107754.2 | NP_001101224.1 | 1 - 2 | 166 | 99 | |
GenBank mRNA | BC168921.1 | - | 1 - 2 | 166 | 9024408 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Rattus norvegicus Probe spans exons |