Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012184.1 | NP_001012184.1 | 9 - 10 | 1336 | 98 | |
XM_006234238.2 | XP_006234300.1 | 9 - 10 | 1093 | 98 | ||
XM_006234241.2 | XP_006234303.1 | 9 - 10 | 1183 | 98 | ||
GenBank mRNA | BC087094.1 | - | 9 - 10 | 1336 | 7191334 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |