Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scrn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013162.1 | NP_001013180.1 | 5 - 6 | 795 | 68 | |
XM_006234358.2 | XP_006234420.1 | 5 - 6 | 832 | 68 | ||
XM_006234359.2 | XP_006234421.1 | 5 - 6 | 834 | 68 | ||
XM_006234360.2 | XP_006234422.1 | 5 - 6 | 834 | 68 | ||
GenBank mRNA | BC088234.1 | - | 5 - 6 | 795 | 7324882 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |