Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fsip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013078.1 | NP_001013096.1 | 2 - 3 | 309 | 65 | |
XM_006234734.2 | XP_006234796.1 | 2 - 3 | 344 | 65 | ||
XM_006234735.2 | XP_006234797.1 | 2 - 3 | 344 | 65 | ||
XM_008762090.1 | XP_008760312.1 | 3 - 4 | 287 | 65 | ||
GenBank mRNA | BC082080.1 | - | 2 - 3 | 309 | 7133293 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |