Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017019.1 | NP_058715.1 | 4 - 5 | 381 | 68 | |
GenBank mRNA | AJ245642.1 | - | 4 - 5 | 345 | 68 | |
D00403.1 | - | 4 - 5 | 381 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |