Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Luzp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030830.2 | NP_110457.2 | 3 - 4 | 257 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |