Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Utp11l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107978.2 | NP_001101448.1 | 3 - 4 | 294 | 123 | |
GenBank mRNA | BC168964.1 | - | 3 - 4 | 294 | 7130611 | 123 |
FQ226285.1 | - | 3 - 4 | 306 | 123 | ||
FQ231750.1 | - | 3 - 4 | 315 | 123 | ||
FQ233930.1 | - | 3 - 4 | 306 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |