Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007651.1 | NP_001007652.1 | 1 - 2 | 38 | 149 | |
GenBank mRNA | BC082028.1 | - | 1 - 2 | 38 | 7130077 | 149 |
FQ211313.1 | - | 1 - 2 | 35 | 149 | ||
FQ211450.1 | - | 1 - 2 | 51 | 149 | ||
FQ219120.1 | - | 1 - 2 | 51 | 149 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |