Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017271.2 | NP_058967.1 | 2 - 3 | 252 | 90 | |
GenBank mRNA | BC065581.1 | - | 2 - 3 | 238 | 5623848 | 90 |
X82445.1 | - | 2 - 3 | 252 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |