Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191818.1 | NP_001178747.1 | 7 - 8 | 768 | 85 | |
XM_008764746.1 | XP_008762968.1 | 8 - 9 | 988 | 85 | ||
XM_008764747.1 | XP_008762969.1 | 9 - 10 | 969 | 85 | ||
XM_008764748.1 | XP_008762970.1 | 8 - 9 | 987 | 85 | ||
XM_008764749.1 | XP_008762971.1 | 7 - 8 | 817 | 85 | ||
XM_008764750.1 | XP_008762972.1 | 8 - 9 | 987 | 85 | ||
XM_008764751.1 | XP_008762973.1 | 7 - 8 | 1234 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Rattus norvegicus Probe spans exons |