Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109477.2 | NP_001102947.2 | 5 - 6 | 405 | 156 | |
XM_006240352.2 | XP_006240414.1 | 6 - 7 | 614 | 156 | ||
GenBank mRNA | BC161941.1 | - | 444 | 156 | ||
BF394767.1 | - | 2 - 3 | 156 | |||
EV765393.1 | - | 6 - 7 | 444 | 156 | ||
FQ211665.1 | - | 409 | 156 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |