Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011984.1 | NP_001011984.1 | 5 - 6 | 810 | 73 | |
XM_006240464.2 | XP_006240526.1 | 3 - 4 | 1455 | 73 | ||
GenBank mRNA | BC085882.1 | - | 5 - 6 | 810 | 7191983 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |