Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057123.1 | NP_476464.1 | 4 - 5 | 484 | 60 | |
GenBank mRNA | BC063157.1 | - | 5 - 6 | 496 | 6919865 | 60 |
D50696.1 | - | 4 - 5 | 484 | 60 | ||
FQ209916.1 | - | 5 - 6 | 489 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |