Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Isca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109278.2 | NP_001102748.1 | 1 - 2 | 72 | 95 | |
GenBank mRNA | FN806632.1 | - | 1 - 2 | 69 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |