Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053511.1 | NP_445963.1 | 5 - 6 | 822 | 86 | |
XM_008764298.1 | XP_008762520.1 | 6 - 7 | 1025 | 86 | ||
GenBank mRNA | AF098301.1 | - | 5 - 6 | 822 | 86 | |
FQ214021.1 | - | 5 - 6 | 815 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |