Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Masp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172043.1 | NP_742040.1 | 7 - 8 | 1048 | 64 | |
XM_008764285.1 | XP_008762507.1 | 7 - 8 | 1104 | 64 | ||
GenBank mRNA | AJ277747.1 | - | 6 - 7 | 990 | 64 | |
BC128700.1 | - | 7 - 8 | 1049 | 7324989 | 64 | |
Y18564.1 | - | 6 - 7 | 827 | 64 | ||
Y18566.1 | - | 4 - 5 | 580 | 64 | ||
Y18571.1 | - | 1 - 2 | 55 | 64 | ||
Y18572.1 | - | 6 - 7 | 885 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |