Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108685.2 | NP_001102155.1 | 4 - 5 | 692 | 69 | |
GenBank mRNA | BC160850.1 | - | 1 - 2 | 165 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |