Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tekt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011977.1 | NP_001011977.1 | 5 - 6 | 637 | 73 | |
XM_006238860.1 | XP_006238922.1 | 5 - 6 | 809 | 73 | ||
XM_006238861.2 | XP_006238923.1 | 4 - 5 | 570 | 73 | ||
GenBank mRNA | BC078990.1 | - | 5 - 6 | 637 | 7112575 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |