Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cela3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106692.1 | NP_001100162.1 | 1 - 2 | 82 | 87 | |
XM_008764220.1 | XP_008762442.1 | 1 - 2 | 90 | 87 | ||
GenBank mRNA | FQ231865.1 | - | 1 - 2 | 62 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Protease Rattus norvegicus Probe spans exons |