Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Necap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006239179.2 | XP_006239241.1 | 2 - 3 | 242 | 59 | |
XM_006239180.2 | XP_006239242.1 | 2 - 3 | 243 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |